ADV390217428 AChemBlock

4-Methylphenylsulfonylurea

  • MDL number MFCD00196522
  • Cas Number : 1694-06-0
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white crystalline powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-methylphenylsulfonylurea.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5806
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart