ADV390217431 AChemBlock

(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride

  • MDL number MFCD04117880
  • Cas Number : 5013-77-4
  • Molecular Weight : 226.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichlorophenyl-n-methylmethanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5816
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart