ADV390217432 AChemBlock

8-Amino-1,3,4,5-tetrahydro-benzo[b]azepin-2-one

  • MDL number MFCD04038431
  • Cas Number : 22246-76-0
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-amino-1-3-4-5-tetrahydro-benzo-b-azepin-2-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5817
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart