ADV390217434 AChemBlock

3-Bromo-5,6,7,8-tetrahydro-imidazo[1,2-a]pyrazine dihydrochloride

  • MDL number MFCD11506085
  • Cas Number : 1057338-30-3
  • Molecular Weight : 274.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-6-7-8-tetrahydro-imidazo-1-2-a-pyrazine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5821
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart