ADV390217445 AChemBlock

tert-butyl 3-(2-oxoethyl)azetidine-1-carboxylate

  • MDL number MFCD18374993
  • Cas Number : 152537-04-7
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-azetidinecarboxylic-acid-3-2-oxoethyl-1-1-dimethylethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5845
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart