ADV390217447 AChemBlock

3-tert-Butoxycarbonylamino-cyclobutane-1,1-dicarboxylic acid diisopropyl ester

  • MDL number MFCD20922763
  • Cas Number : 1363382-20-0
  • Molecular Weight : 343.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-tert-butoxycarbonylamino-cyclobutane-1-1-dicarboxylic-acid-diisopropyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5849
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart