ADV390217449 AChemBlock

1,2,3-Oxathiazolidine,3-ethyl-,2,2-dioxide

  • MDL number MFCD20040498
  • Cas Number : 153631-34-6
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-2-3-oxathiazolidine-3-ethyl-2-2-dioxide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5852
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart