ADV390217450 AChemBlock

Ethyl 4-aminomethyl-1-Boc-piperidine-4-carboxylate

  • MDL number MFCD14525454
  • Cas Number : 1016258-69-7
  • Molecular Weight : 286.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-aminomethyl-1-boc-piperidine-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5854
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart