ADV390217452 AChemBlock

3-Amino-oxetane-3-carboxylic acid methyl ester

  • MDL number MFCD20922764
  • Cas Number : 1363383-31-6
  • Molecular Weight : 131.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-amino-oxetane-3-carboxylic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5856
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart