ADV390217456 AChemBlock

N-[3-[(5-Chloro-1H-pyrrolo[2,3-b]pyridin-3-yl)carbonyl]-2,4-difluorophenyl]-1-propanesulfonamide

  • MDL number MFCD14635203
  • Cas Number : 918505-84-7
  • Molecular Weight : 413.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-3-5-chloro-1h-pyrrolo-2-3-b-pyridin-3-yl-carbonyl-2-4-difluorophenyl-1-propanesulfonamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5861
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart