ADV390217464 AChemBlock

2-(4-oxo-3,4-dihydrophthalazin-1-yl)acetic acid

  • MDL number MFCD00085017
  • Cas Number : 25947-11-9
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-phthalazineacetic-acid-3-4-dihydro-4-oxo.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5874
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart