ADV390217465 AChemBlock

6-(2-Bromoacetyl)-3,4-dihydro-1H-quinoline-2-one

  • MDL number MFCD00173687
  • Cas Number : 70639-82-6
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form rownish solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-2-bromoacetyl-3-4-dihydro-1h-quinoline-2-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5875
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart