ADV390217469 AChemBlock

1-Chloro-[2,7]naphthyridine

  • MDL number MFCD00234389
  • Cas Number : 69042-30-4
  • Molecular Weight : 164.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-chloro-2-7-naphthyridine.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3441
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart