ADV390217478 AChemBlock

3-Methyl-3-aminomethylazetidine dihydrochloride

  • MDL number MFCD09264501
  • Cas Number : 96308-73-5
  • Molecular Weight : 173.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-3-aminomethylazetidine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5893
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart