ADV390217481 AChemBlock

ethyl 2,6-dichloro-5-cyanonicotinate

  • MDL number MFCD22415252
  • Cas Number : 919354-52-2
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-pyridinecarboxylic-acid-2-6-dichloro-5-cyano-ethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5897
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart