ADV390217482 AChemBlock

5-Bromo-6-fluoro-3-methyl-1H-indazole

  • MDL number MFCD11044590
  • Cas Number : 864773-66-0
  • Molecular Weight : 229.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-fluoro-3-methyl-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5898
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart