ADV390217484 AChemBlock

5-Amino-6-methyl-3-pyridinecarboxylic acid ethyl ester

  • MDL number MFCD18256589
  • Cas Number : 1008138-73-5
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-6-methyl-3-pyridinecarboxylic-acid-ethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5900
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart