ADV390217545 AChemBlock

Methyl 3-Boc-3-azabicyclo[3.1.1]heptane-6-carboxylate

  • MDL number MFCD21642061
  • Cas Number : 1363380-75-9
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-boc-3-azabicyclo-3-1-1-heptane-6-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6133
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart