ADV390217572 AChemBlock

trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride

  • MDL number MFCD23105899
  • Cas Number : 2006333-41-9
  • Molecular Weight : 141.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/trans-4-fluoro-3-hydroxypyrrolidine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6498
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart