ADV390217577 AChemBlock

5-Bromopyrrolo[2,1-f][1,2,4]triazin-4(1H)-one

  • MDL number MFCD23703616
  • Cas Number : 1403767-05-4
  • Molecular Weight : 214.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromopyrrolo-2-1-f-1-2-4-triazin-4-1h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6703
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart