ADV390217588 AChemBlock

4-(4,4-Difluoro-piperidin-1-yl)-phenylamine

  • MDL number MFCD23163472
  • Cas Number : 1266330-18-0
  • Molecular Weight : 212.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-4-4-difluoro-piperidin-1-yl-phenylamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4481
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart