ADV390217597 AChemBlock

5-(3-Azetidinyl)-2H-tetrazole hydrochloride

  • MDL number MFCD21647389
  • Cas Number : 950691-49-3
  • Molecular Weight : 161.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-3-azetidinyl-2h-tetrazole-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5993
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart