ADV390217602 AChemBlock

7-Chloro-4-methoxy-1(2H)-isoquinolinone

  • MDL number MFCD20922768
  • Cas Number : 630423-35-7
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-4-methoxy-1-2h-isoquinolinone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6001
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart