ADV390217619 AChemBlock

5-Methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00090754
  • Cas Number : 402-61-9
  • Molecular Weight : 126.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white to light-yellow powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1h-pyrazole-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6037
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart