ADV390217633 AChemBlock

1-Amino-3,3-difluoro-cyclobutanecarboxylic acid

  • MDL number MFCD17480490
  • Cas Number : 1225532-86-4
  • Molecular Weight : 151.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-amino-3-3-difluoro-cyclobutanecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6066
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart