ADV390217640 AChemBlock

Dimethyl 3-amino-1,2-benzenedicarboxylate

  • MDL number MFCD09029586
  • Cas Number : 34529-06-1
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-3-amino-1-2-benzenedicarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6076
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart