ADV390217647 AChemBlock

3-Bromo-7-nitro-1H-indazole

  • MDL number MFCD00159910
  • Cas Number : 74209-34-0
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pale-yellow powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-7-nitro-1h-indazole.html

Safety Information

signalword Danger
pcodes P201;P202;P281;P308+313
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6091
pictograms GHS08
hstatements H360
Availability details for
Quantity
  • Availability
  • Add to Cart