ADV390217653 AChemBlock

oxazol-4-ylmethanamine hydrochloride

  • MDL number MFCD08437648
  • Cas Number : 1072806-60-0
  • Molecular Weight : 134.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-oxazolemethanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6099
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart