ADV390217661 AChemBlock

(R)-tert-Butyl 2-(pyrrolidin-2-yl)ethylcarbamate

  • MDL number MFCD06656825
  • Cas Number : 720000-05-5
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-tert-butyl-2-pyrrolidin-2-yl-ethylcarbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6112
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart