ADV390217665 AChemBlock

Methyl trans-3-(Boc-amino)cyclobutanecarboxylate

  • MDL number MFCD20922912
  • Cas Number : 1101173-77-6
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-trans-3-boc-amino-cyclobutanecarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6121
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart