ADV390217681 AChemBlock

Ethyl 3-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD01006747
  • Cas Number : 3284-47-7
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white crystals
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-methyl-1h-pyrrole-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6138
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart