ADV390217696 AChemBlock

7-Boc-2-cyano-7-azaspiro[3.5]nonane

  • MDL number MFCD12198800
  • Cas Number : 203662-66-2
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/7-boc-2-cyano-7-azaspiro-3-5-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6166
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart