ADV390217704 AChemBlock

2-Amino-7-Cbz-7-azaspiro[3.5]nonane

  • MDL number MFCD15071709
  • Cas Number : 147611-02-7
  • Molecular Weight : 274.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-7-cbz-7-azaspiro-3-5-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6175
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart