ADV390217728 AChemBlock

9-Benzyl-3-oxa-9-azabicyclo[3.3.1]nonan-7-one

  • MDL number MFCD20134206
  • Cas Number : 81514-40-1
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/9-benzyl-3-oxa-9-azabicyclo-3-3-1-nonan-7-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6211
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart