ADV390217730 AChemBlock

6,8-Dibromoimidazo[1,2-a]pyrazine

  • MDL number MFCD08460056
  • Cas Number : 63744-22-9
  • Molecular Weight : 276.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form beige solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-8-dibromoimidazo-1-2-a-pyrazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6213
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart