ADV390217734 AChemBlock

6,6-Dimethyl-3-azabicyclo[3.1.0]hexane hydrochloride

  • MDL number MFCD20441578
  • Cas Number : 943516-55-0
  • Molecular Weight : 147.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-6-dimethyl-3-azabicyclo-3-1-0-hexane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6218
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart