ADV390217735 AChemBlock

2-Formyl-1-trityl-aziridine

  • MDL number MFCD20926171
  • Cas Number : 173277-15-1
  • Molecular Weight : 313.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-formyl-1-trityl-aziridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6219
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart