ADV390217745 AChemBlock

1-Boc-3-methylpyrrolidine-3-methanol

  • MDL number MFCD20926173
  • Cas Number : 1263506-20-2
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-methylpyrrolidine-3-methanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6230
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart