ADV390217747 AChemBlock

3-(4-Chloro-phenyl)-pyrazine-2-carboxylic acid methyl ester

  • MDL number MFCD20926174
  • Cas Number : 1363382-34-6
  • Molecular Weight : 248.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-chloro-phenyl-pyrazine-2-carboxylic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6232
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart