ADV390217761 AChemBlock

4-Amino-3-iodo-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD22566154
  • Cas Number : 862729-12-2
  • Molecular Weight : 275.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-3-iodo-1-methyl-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6255
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart