ADV390217765 AChemBlock

2-Boc-5-oxo-hexahydrocyclopenta[c]pyrrole

  • MDL number MFCD11616063
  • Cas Number : 146231-54-1
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-5-oxo-hexahydrocyclopenta-c-pyrrole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6259
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart