ADV390217767 AChemBlock

3-Hydroxy-3-(trifluoromethyl)-cyclobutanecarboxylic acid

  • MDL number MFCD11870383
  • Cas Number : 1163729-49-4
  • Molecular Weight : 184.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxy-3-trifluoromethyl-cyclobutanecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6261
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart