ADV390217780 AChemBlock

1-Boc-3-bromoazetidine

  • MDL number MFCD16658899
  • Cas Number : 1064194-10-0
  • Molecular Weight : 236.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-bromoazetidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6278
pictograms GHS05; GHS07
hstatements H302;H313;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart