ADV390217781 AChemBlock

1-Diphenylmethyl-3-methyleneazetidine

  • MDL number MFCD11559000
  • Cas Number : 40569-55-9
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-diphenylmethyl-3-methyleneazetidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6279
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart