ADV390217784 AChemBlock

1-Boc-3-(hydroxymethyl)-3-methylazetidine

  • MDL number MFCD20926177
  • Cas Number : 1363382-91-5
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-hydroxymethyl-3-methylazetidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6282
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart