ADV390217790 AChemBlock

2-(1H-1,2,3-Triazol-1-yl)benzoic acid

  • MDL number MFCD20486487
  • Cas Number : 1085458-53-2
  • Molecular Weight : 189.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-1h-1-2-3-triazol-1-yl-benzoic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6289
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart