ADV390217795 AChemBlock

3-Aminomethyl-3-(hydroxymethyl)oxetane

  • MDL number MFCD09800579
  • Cas Number : 45513-32-4
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-aminomethyl-3-hydroxymethyl-oxetane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6296
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart