ADV390217802 AChemBlock

Diisopropyl 3,3-difluorocyclobutane-1,1-dicarboxylate

  • MDL number MFCD23105827
  • Cas Number : 1225532-89-7
  • Molecular Weight : 264.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/diisopropyl-3-3-difluorocyclobutane-1-1-dicarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6306
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart