ADV390217813 AChemBlock

2-Acetylpyrimidine

  • MDL number MFCD08361636
  • Cas Number : 53342-27-1
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-acetylpyrimidine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6326
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart