ADV390217836 AChemBlock

4-Amino-4-hydroxymethyl-tetrahydro-2H-pyran

  • MDL number MFCD04115578
  • Cas Number : 720706-20-7
  • Molecular Weight : 131.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-4-hydroxymethyl-tetrahydro-2h-pyran.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6353
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart